missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP1 Antibody (OTI4H1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74487G
This item is not returnable.
View return policy
Description
TDP1 Monoclonal specifically detects TDP1 in Human samples. It is validated for Western Blot.
Specifications
| TDP1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TDP1 | |
| Full length recombinant protein of human TDP1 (NP_060789) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI4H1 | |
| Western Blot | |
| EC 3.1.4, EC 3.1.4.-, FLJ11090, MGC104252, SCAN1, Tyr-DNA phosphodiesterase 1, tyrosyl-DNA phosphodiesterase 1 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 55775.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction