missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567165
This item is not returnable.
View return policy
Description
Immunogen sequence: STSSLLCARQG AANEPRYTCS EAQKAAHKRK ISPVKFSNTD SVLPPKRQKS GSQEDLGWCL SSSDDELQPE M Highest antigen sequence identity to the following orthologs - mouse 65%, rat 72%.
The protein encoded by this gene is involved in repairing stalled topoisomerase I-DNA complexes by catalyzing the hydrolysis of the phosphodiester bond between the tyrosine residue of topoisomerase I and the 3-prime phosphate of DNA. This protein may also remove glycolate from single-stranded DNA containing 3-prime phosphoglycolate, suggesting a role in repair of free-radical mediated DNA double-strand breaks. This gene is a member of the phospholipase D family and contains two PLD phosphodiesterase domains. Mutations in this gene are associated with the disease spinocerebellar ataxia with axonal neuropathy (SCAN1). While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
Specifications
| TDP1 | |
| Polyclonal | |
| Unconjugated | |
| Tdp1 | |
| 2810481F14Rik; 4921509N21Rik; AI838772; AW493413; E430034L06Rik; MLZ-501; protein expressed in male leptotene and zygotene spermatocytes 501; SCAN1; Tdp1; Tyr-DNA phosphodiesterase 1; tyrosyl-DNA phodphodiesterase 1; tyrosyl-DNA phosphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUW8 | |
| Tdp1 | |
| Recombinant Human TDP1. Recombinant protein control fragment (Product #RP-105529). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction