missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588834
This item is not returnable.
View return policy
Description
Immunogen sequence: PFNFEKNENK LPPHESLRSP GTLPNHPNFR LKSSENGNKK NNFLLCEQTK QYLASQEDNS VSSNPNGING EVVGSKGDRK KLPAGNSVSP PSAESNSPPK E; Positive Samples: Mouse testis; Cellular Location: Cytoplasm.
This gene is similar to a mouse gene that encodes a tudor domain protein. Alternatively spliced transcript variants have been described but their full length sequences have not been determined.
Specifications
| TDRD1 | |
| Polyclonal | |
| Unconjugated | |
| Tdrd1 | |
| Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83561 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99MV1 | |
| Tdrd1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-125 of human TDRD1 (NP_9420901). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction