missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD3 Antibody (RAB-C419) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
€ 399.00 - € 849.00
Specifications
| Antigen | TDRD3 |
|---|---|
| Clone | RAB-C419 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660294
|
Novus Biologicals
NBP3-20148-0.1mg |
0.1 mg |
€ 849.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602195
|
Novus Biologicals
NBP3-20148-0.025mg |
0.025 mg |
€ 399.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TDRD3 Monoclonal antibody specifically detects TDRD3 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| TDRD3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ21007, tudor domain containing 3, tudor domain-containing protein 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing TDRD3 protein under non-denaturing conditions. | |
| Affinity purified |
| RAB-C419 | |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| Unconjugated | |
| Human | |
| Neurodegeneration, Neuroscience | |
| PBS | |
| 81550 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title