missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRD3 Recombinant Human Monoclonal Antibody (RAB-C419)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551839
This item is not returnable.
View return policy
Description
This antibody recognizes TDRD3 (Tudor domain-containing protein 3). It binds to a folded domain, amino acids 1-455. TDRD3 is a scaffolding protein that specifically recognizes and binds dimethylarginine-containing proteins.
TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predominantly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing variants. In humans, the gene is located on chromosome 13.
Specifications
| TDRD3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9H7E2 | |
| TDRD3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing TDRD3 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Immunocytochemistry, Mass Spectrometry | |
| RAB-C419 | |
| Unconjugated | |
| TDRD3 | |
| 4732418C03Rik; 6720468N18; Tdrd3; tudor domain containing 3; tudor domain-containing protein 3 | |
| Human | |
| Metal-chelate Chromatography | |
| RUO | |
| 81550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction