missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | TDRD9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TDRD9 Polyclonal antibody specifically detects TDRD9 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| TDRD9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| C14orf75, chromosome 14 open reading frame 75, DKFZp434N0820, EC 3.6.1, EC 3.6.4.13, FLJ36164, HIG-1, hypoxia-inducible HIG-1, MGC135025, NET54, putative ATP-dependent RNA helicase TDRD9, tudor domain containing 9, Tudor domain-containing protein 9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human TDRD9 (NP_694591.2). AVPVQNKMNPAYIFEVEGKPHSVEEYYLNDLEHIHHSKLSPHLLEEPVITKDIYEVAVSLIQMFDDLDMKESGNKAWSGAQFVLERSSVLVFLPGLGEINYMHELLTSLVHKRLQVYPLHSSVALEEQNNVFLSPVPGYRKIILSTNIAESSVTVPDVKYVIDFCLTRTLVCDEDTNYQSLRLSWASKTSCNQRKGRAGRVSRGYCYRLVHKDFWDNSIPDHVVPEMLRCPLGSTILKVKLLDMGEPRALLATALSPPGLSDIERTILLLKEVGALAVSGQREDENPHDGELTFLGRVLAQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 122402 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title