missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568740
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 77%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%.
TDRD9 is a protein coding gene.
Specifications
| TDRD9 | |
| Polyclonal | |
| Unconjugated | |
| TDRD9 | |
| 4930441E05Rik; ATP-dependent RNA helicase TDRD9; C14orf75; HIG-1; hypoxia-inducible HIG-1; NET54; putative ATP-dependent RNA helicase TDRD9; TDRD9; tudor domain containing 9; tudor domain-containing protein 9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 122402 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NDG6 | |
| TDRD9 | |
| synthetic peptide directed towards the middle region of human TDRD9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction