missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRKH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13528-1-AP-150UL
This item is not returnable.
View return policy
Description
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.Specifications
| TDRKH | |
| Polyclonal | |
| Unconjugated | |
| TDRKH | |
| TDRD2, TDRKH, tudor and KH domain containing | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11022, 310652, 72634 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80VL1, Q9Y2W6 | |
| Tdrkh | |
| TDRKH specific Fusion Protein Ag4422 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction