missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570994
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 79%; Rat: 100%.
Contributes to normal sperm motility, but not essential for male fertility.
Specifications
| TDRP | |
| Polyclonal | |
| Unconjugated | |
| TDRP | |
| 2610019F03Rik; C25H8orf42; C8orf42; Inm01; Protein INM01; TDRP; TDRP1; TDRP2; testis development related protein; testis development-related protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 157695 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86YL5 | |
| TDRP | |
| synthetic peptide directed towards the N-terminal region of human TDRP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction