missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Antibody (DNTT/4506R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08564FR
This item is not returnable.
View return policy
Description
TdT Monoclonal antibody specifically detects TdT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TdT | |
| Monoclonal | |
| DyLight 680 | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| Synthetic peptide corresponding to residues within aa1-100 of TdT was used as an immunogen. (Uniprot: P04053) | |
| 100 μg | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 1791 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DNTT/4506R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction