missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Antibody (rDNTT/6909), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14198PE
This item is not returnable.
View return policy
Description
TdT Monoclonal antibody specifically detects TdT in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| TdT | |
| Monoclonal | |
| PE | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| Recombinant fragment (around aa1-100) of human TdT protein (exact sequence is proprietary) (Uniprot: P04053) | |
| 0.1 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 1791 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| rDNTT/6909 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction