missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEAD2 Polyclonal Antibody

Description
Immunogen sequence: TARLQLVEFSA FVEPPDAVDS YQRHLFVHIS QHCPSPGAPP LESVDV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 96%.
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5'-GTGGAATGT-3'). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.
Specifications
Specifications
| Antigen | TEAD2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TEAD2 |
| Gene Accession No. | Q15562 |
| Gene Alias | Embryonic TEA domain containing factor; embryonic TEA domain-containing factor; Etdf; ETEF-1; Etf; TEA domain family member 2; TEA domain transcription factor 2; Tead2; TEAD-2; TEF4; TEF-4; transcriptional enhancer factor TEF-4 |
| Gene Symbols | TEAD2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?