Learn More
TEAD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83949
This item is not returnable.
View return policy
Description
TEAD3 Polyclonal specifically detects TEAD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| 0.1 mL | |
| IgG |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QDIKPFAQPAYPIQPPLPPTLSSYEPLAPLPSAAASVPVWQDRTIASSRLRLLEYSAFMEVQRDPDTYSKHLFVHIGQTNPAFSDPPLEAVDVRQIYDKFPEK | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |