missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEB4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518868
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
MARCH6 is an E3 ubiquitin ligase that localizes to endoplasmic reticulum membranes.
Specifications
| TEB4 | |
| Polyclonal | |
| Unconjugated | |
| MARCHF6 | |
| 3830408G03; 6-Mar; DOA10; doa10 homolog; E3 ubiquitin-protein ligase MARCH6; F830029L24Rik; KIAA0597; MARCH6; MARCH-VI; membrane associated ring finger 6; membrane associated ring-CH-type finger 6; membrane-associated ring finger (C3HC4) 6; membrane-associated ring finger (C3HC4) 6, E3 ubiquitin protein ligase; membrane-associated RING finger protein 6; membrane-associated RING-CH finger protein 6; membrane-associated RING-CH protein VI; mKIAA0597; Protein TEB-4; RING finger protein 176; RING-type E3 ubiquitin transferase MARCH6; RNF176; TEB4 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 10299 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60337 | |
| MARCHF6 | |
| Synthetic peptide sequence (QGRRVIFQKVKE) corresponding to the internal amino acids of 38777 (aa 701-713). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction