Learn More
Techne™ Replacement Block
Product Code. 12648875
Change view
Click to view available options
Capacity:
18 x 0.5 mL
24 x 0.2 mL
30 x 0.5 mL
48 x 0.2 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12648875 18 x 0.5 mL Each
12628875 24 x 0.2 mL Each
12688875 48 x 0.2 mL Each
12608885 30 x 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12648875 Supplier Techne™ Supplier No. 9995799
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Parts and accessories to keep your Techne™ 3 Prime thermal cycler working optimally.
Specifications
For Use With (Equipment) Thermal Cycler
Capacity 18 x 0.5 mL
Product Type Replacement Block