Learn More
Cole-Parmer™ Adjustable Bath Tray for Cole-Parmer™ IC-300 Series Circulators and WBU-200 Series Water Baths
Click to view available options
Capacity (Metric):
8 L
12 L
18 L
26 L
48 L
For Use With (Equipment):
Unheated water bath 16101-88
Unheated water bath 16101-89
Unheated water bath 16104-45
Unheated water bath 16104-53
Unheated water bath 16112-01
Unit Size:
Each
Description
- Lids, trays, temperature probes, and other accessories for your bath.
Specifications
Specifications
| Description | Adjustable Bath Tray, for 8 Liter Baths |
| Capacity (Metric) | 8 L |
| For Use With (Equipment) | Unheated water bath 16104-53 |
| Type | Adjustable Tray |
| Material | Stainless Steel |