Learn More
Hettich Lab Technology™ Manual Hand Centrifuge
Description
- Self-lubricating drive system makes it virtually maintenance-free
- Does not require electricity - can run in place like remote pastures or in case of electrical failure
- Excellent for the veterinary industry, field testing, aid organizations and pharmacies
Specifications
Specifications
| Capacity | 4 x 15 mL |
| Max. RCF | 1,298 x g |
| Max. Speed | 3,000 rpm |
| Refrigerated | No |
| Drive System | Self-lubricating |
| Rotor | Rotor must be purchased separately (1014, 1025). |
| Type | Hand Centrifuge |
| Dimensions (HWD) | 285 x 140 x 175 mm |
| Net Weight | 0.9 kg |