Learn More
Usbeck Laborgeraete™ Teclu Burner
Click to view available options
Gas Type:
Natural Gas
Propane
Unit Size:
Each
Description
- Air regulation
- Needle valve
- Variant of Bunsen burner
Specifications
Specifications
| Product Type | Teclu Burner |
| Gas Type | Propane |
| Diameter (Metric) | 17 mm (Head) |
| Diameter (Metric) Outer Mixing Tube | 14 mm |
| Height (Metric) | 155 mm |
| Max. Temperature (Metric) | 1530°C |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 260 g |