Learn More
Saint Gobain Life Sciences Chemware™ PFA Sieves
Product Code. 10460965
Change view
Click to view available options
Pore Size:
1 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10460965 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10460965 Supplier Saint Gobain Life Sciences Supplier No. D1069101
Only null left
Add to basket
Edge
Add to basket
Chemware™ PFA Sieves are resistant to most chemicals and are ideal for ultra-pure applications.
  • Non-contaminating and resistant to most chemicals
  • Ideal for coarse sorting or draining of aggressive media
  • Handle hooks over edge of vessel for draining
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 1 mm
Type Sieve
Diameter (English) Outer 3 in.
Material PFA
U.S. Standard No. 16
Size (Metric) 180 mm long with handle
Diameter (Metric) Outer 76 mm
Screen Size 16 Mesh
Sieve Number 16