missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teicoplanin, MedChemExpress
Description
Teicoplanin is a glycopeptide antibiotic used in the prophylaxis and treatment of serious infections caused by Gram-positive bacteria, including Methicillin-resistant Staphylococcus aureus and Enterococcus faecalis.
Specifications
Specifications
| Chemical Name or Material | Teicoplanin |
| Physical Form | Powder |
| CAS | 61036-62-2 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Antibiotic MDL-507 MDL-507 |
| Solubility Information | H2O : 100 mg/mL (Need ultrasonic) ∣DMSO : 100 mg/mL (Need ultrasonic) |
| SMILES | ClC1=C(OC2=CC3=CC(OC4=CC=C5C=C4Cl)=C2O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6N[R])C=CC([C@H]([C@@H]7NC([C@](C8=CC(C9=C([C@H](C(O)=O)NC7=O)C=C(O)C=C9O[C@@H]%10[C@@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O%10)=C(O)C=C8)([H])NC([C@]3([H])NC([C@@](C%11=CC(O)=CC(OC%12=C(O)C=CC([C@@H](N)C%13=O)=C%12)=C%11)([H])NC([C@H](N%13)C5)=O)=O)=O)=O)O[C@@H]%14O[C@H](CO)[C@@H](O)[C@H](O)[C@H]%14NC(C)=O)=C1.CCCCC/C=C/CCC([R])=O.CC(C)CCCCCCC([R])=O.CCCCCCCCCC([R])=O.CCC(C)CCCCCCC([R])=O.CC(C)CCCCCCCC([R])=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?