Learn More
Invitrogen™ TEKT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140263
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: WYRSNLTNYQ ESNTSRHNSE KLRVDTSRLI QDKYQQTRKT QADTTQNLGE RVNDIGFWKS EIIHELDEMI GETNALTDVK KRLERALMET EAP Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 83%.
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
Specifications
| TEKT3 | |
| Polyclonal | |
| Unconjugated | |
| TEKT3 | |
| 4933407G07Rik; OMC45; TEKT3; tektin 3; tektin-3; testicular microtubules-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64518 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXF9 | |
| TEKT3 | |
| Recombinant protein corresponding to Human TEKT3. Recombinant protein control fragment (Product #RP-109358). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.