missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT5 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81832F
This item is not returnable.
View return policy
Description
TEKT5 Polyclonal antibody specifically detects TEKT5 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TEKT5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| FLJ32871, MGC163208, MGC163210, tektin 5, tektin-5 | |
| TEKT5 antibody was raised against a 17 amino acid synthetic peptide near the center of human TEKT5. The immunogen is located within amino acids 220 - 270 of TEKT5. Amino Acid Squence: DIQMRDNRDAQHVLERD | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 146279 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction