missing translation for 'onlineSavingsMsg'
Learn More

TEL2 Antibody (8B6) [FITC], Novus Biologicals Biologicals™

Product Code. 30522264 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522264 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522264 Supplier Novus Biologicals Supplier No. NBP343043F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEL2 Monoclonal antibody specifically detects TEL2 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEL2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 8B6
Conjugate FITC
Formulation PBS
Gene Alias CLK2, DKFZp434A073, EC 1.1.1.27, EC 2.7.11.1, HCLK2, KIAA0683FLJ10924, Protein clk-2 homolog, TEL2, TEL2, telomere maintenance 2, homolog (S. cerevisiae), telomere length regulation protein TEL2 homolog
Host Species Mouse
Immunogen TEL2, telomere maintenance 2, homolog(S. cerevisiae) - (Uniprot# Q9Y4R8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 9894
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.