missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telbivudine, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Telbivudine |
| CAS | 3424-98-4 |
| Molecular Formula | C10 H14 N2 O5 |
| Quantity | 5 mg |
| Synonym | 2,4(1H,3H)-Pyrimidinedione','1-(2-deoxy-β-L-erythro-pentofuranosyl)-5-methyl-, Thymine','1-(2-deoxy-β-L-erythro-pentofuranosyl)- (7CI,8CI), 1-(2-Deoxy-β-L-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione, 2'-Deoxy-L-thymidine, Epavudine, L-Deoxythymidine, L-Thymidine, NV 02B, Sebivo, Telbivudine, Tyzeka, β-L-Thymidine |
| InChI Formula | InChI=1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m1/s1 |
| SMILES | CC1=CN([C@@H]2C[C@@H](O)[C@H](CO)O2)C(=O)NC1=O |
| IUPAC Name | 1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 242.23 |
| Formula Weight | 242.0903 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction