Learn More
MedChemExpress Telbivudine
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Molecular Weight (g/mol) | 242.23 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.05% |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (206.42 mM∣Need ultrasonic) ∣H2O : 33.33 mg/mL (137.60 mM∣Need ultrasonic) |
| For Use With (Application) | COVID-19-anti-virus |
| CAS | 3424-98-4 |
| Synonym | Epavudine∣ L-Thymidine∣ NV 02B |