missing translation for 'onlineSavingsMsg'
Learn More

TELO2 Mouse anti-Human, Rat, Clone: 3H10F4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66077-1-IG-150UL

 View more versions of this product

Product Code. 16847625

  • € 410.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.
Specifications

Specifications

TELO2
Monoclonal
1.68 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y4R8
Telo2
TELO2 Fusion Protein Ag8834
150 μL
Primary
Human, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
3H10F4
Unconjugated
TELO2
hCLK2, KIAA0683, Protein clk 2 homolog, TEL2, TELO2
Mouse
Protein G
RUO
302986, 9894
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.