missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM5/GPR124 Antibody (499716) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8896JF635
This item is not returnable.
View return policy
Description
TEM5/GPR124 Monoclonal antibody specifically detects TEM5/GPR124 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| TEM5/GPR124 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DKFZp434C211, DKFZp434J0911, G protein-coupled receptor 124, G-protein coupled receptor 124, KIAA1531FLJ14390, TEM5Tumor endothelial marker 5 | |
| Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124, Met1-Leu699, Accession # Q96PE1 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 499716 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Endothelial Cell Markers, GPCR | |
| 25960 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction