missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM7/PLXDC1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97892UV
This item is not returnable.
View return policy
Description
TEM7/PLXDC1 Polyclonal specifically detects TEM7/PLXDC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TEM7/PLXDC1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
| Produced in rabbits immunized with E. coli-derived Human TEM7/PLXDC1 fragment. | |
| 0.1 mL | |
| Angiogenesis | |
| 57125 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction