missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM8/ANTXR1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21501AF532
This item is not returnable.
View return policy
Description
TEM8/ANTXR1 Polyclonal antibody specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TEM8/ANTXR1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8 | |
| Antibody was raised against a peptide corresponding to 13 amino acids near the C-terminus of human TEM8/ANTXR1. The immunogen is located within the last 50 amino acids.. Amino Acid Squence: RAPPPSRPPPRPSV | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific | |
| 84168 | |
| Store at 4C in the dark. |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction