Learn More
Horst™ Temperature Controllers
Click to view available options
Model:
HT 30
HT 31
HT30
HT31
Sensor Types:
NiCr-Ni (K)
Pt 100
Unit Size:
Each
Specifications
Specifications
| Wattage | 2200 or 3500w |
| Power Density | 230 V - 50 Hz |
| Model | HT30 |
| Sensor Types | NiCr-Ni (K) |
| Amperage | 10 A |
| Type | Temperature Controller |
| Voltage | 230 V |