Learn More
Binder™ Temperature and Humidity Measurement
Product Code. 17872445 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
Binder™ Walk-in Chamber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17872445 Binder™ Walk-in Chamber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17872445 Supplier Binder™ Supplier No. DL300113
Temperature measurement service of temperature and humidity measuring points, as well as certificate.
  • Service includes 13 temperature measuring points and 1 humidity measuring point in center of walk-in chamber with a pair of values specified by the user
  • Includes certificate
Specifications
For Use With (Equipment) Binder™ Walk-in Chamber
Product Type Measurement Service