Learn More
Cole-Parmer™ Temperature Probe for Cole-Parmer™ Stuart Stirring Hot Plates and Hot Plates
Description
- Probe for Temperature Controller.
- Accurate liquid temperature control.
Specifications
Specifications
| For Use With (Equipment) | SCT1 Temperature Controller |
| Type | Temperature Probe |
| Material | PTFE |