Learn More
Memmert™ Temperature Restriction for IF750
Product Code. 17567155 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF750 Incubator
Unit Size:
Each
Product Code. 17567155
Brand: Memmert™ IF750A8
Only null left
Add to basket
Add to basket
Optional temperature restriction between 50°C and 75°C.
  • Accessory
  • 1 Choice
  • Temperatures: 50°C, 55°C, 60°C, 65°C, 70°C, or 75°C
Specifications
For Use With (Equipment) IF750 Incubator
Type Temperature Restriction