Learn More
Memmert™ Temperature Restriction for IN30
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN30 Incubator
Unit Size:
Each
Description
- Accessory
- 1 Choice
- Temperatures: 50°C, 55°C, 60°C, 65°C, 70°C, or 75°C
Specifications
Specifications
| For Use With (Equipment) | IN30 Incubator |
| Type | Temperature Restriction |