missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenapanor, MedChemExpress
Description
Tenapanor (AZD1722) is a potent and orally active sodium/hydrogen exchanger isoform 3 (NHE3) inhibitor. Tenapanor reduces intestinal phosphate absorption predominantly through reduction of passive paracellular phosphate flux. Tenapanor has the potential for the research of hyperphosphatemia.
Specifications
Specifications
| Chemical Name or Material | Tenapanor |
| CAS | 1234423-95-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C50H66Cl4N8O10S2 |
| Quantity | 50 mg |
| Synonym | AZD1722 RDX5791 |
| Solubility Information | DMSO : 50 mg/mL (43.67 mM; Need ultrasonic) |
| SMILES | O=C(NCCOCCOCCNS(=O)(C1=CC=CC([C@@H]2CN(C)CC3=C2C=C(Cl)C=C3Cl)=C1)=O)NCCCCNC(NCCOCCOCCNS(=O)(C4=CC=CC([C@@H]5CN(C)CC6=C5C=C(Cl)C=C6Cl)=C4)=O)=O |
| Molecular Weight (g/mol) | 1145.05 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?