missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin C Antibody (4C8MS), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68136MFV450
This item is not returnable.
View return policy
Description
Tenascin C Monoclonal antibody specifically detects Tenascin C in Human, Mouse, Rat, Feline samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| Tenascin C | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Feline | |
| Purified |
| Flow Cytometry | |
| 4C8MS | |
| Flow Cytometry, Flow (Intracellular) | |
| 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
| Recombinant human Tenascin C [Swiss-Prot# P24821]. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Prostate Cancer | |
| 3371 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction