missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin C Antibody (TNC/2981R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08946UV
This item is not returnable.
View return policy
Description
Tenascin C Monoclonal antibody specifically detects Tenascin C in Human, Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Tenascin C | |
| Monoclonal | |
| DyLight 350 | |
| 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
| Recombinant human full-length Tenascin C protein (Uniprot: P24821) | |
| 100 μg | |
| Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer | |
| 3371 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| TNC/2981R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction