missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teneurin-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41315
This item is not returnable.
View return policy
Description
Teneurin-1 Polyclonal specifically detects Teneurin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Teneurin-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TENM1 | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of human TENM1. The immunogen is located within the first 50 amino acids of TENM1. Amino Acid Squence: SDESEDGRKPRQSFNSRE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| TENM1 antibody is predicted to not cross-react with other members of the TENM family. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 - 2 ug/mL, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
| odz, odd Oz/ten-m homolog 1(Drosophila), Protein Odd Oz/ten-m homolog 1, ten-1, tenascin M, tenascin M1, tenascin-M1, teneurin 1, teneurin-1, Ten-m1, TNM, TNM1 | |
| Rabbit | |
| 280 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 10178 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction