missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teneurin-1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41315R
This item is not returnable.
View return policy
Description
Teneurin-1 Polyclonal antibody specifically detects Teneurin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Teneurin-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| odz, odd Oz/ten-m homolog 1(Drosophila), Protein Odd Oz/ten-m homolog 1, ten-1, tenascin M, tenascin M1, tenascin-M1, teneurin 1, teneurin-1, Ten-m1, TNM, TNM1 | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of human TENM1. The immunogen is located within the first 50 amino acids of TENM1. Amino Acid Squence: SDESEDGRKPRQSFNSRE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10178 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction