missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir alafenamide, MedChemExpress

Product Code. 16486676
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486676

Brand: Medchem Express HY15232

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tenofovir alafenamide (GS-7340) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.

Tenofovir alafenamide (GS-7340) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.

Specifications

Chemical Name or Material Tenofovir alafenamide
CAS 379270-37-8
Color White
Purity Grade Notes Research
Molecular Formula C21H29N6O5P
Quantity 100 mg
Synonym GS-7340
Solubility Information DMSO : ≥ 31 mg/mL (65.06 mM) ∣H2O : 6.67 mg/mL (14.00 mM; Need ultrasonic)
SMILES NC1=NC=NC2=C1N=CN2C[C@@H](C)OC[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O
Molecular Weight (g/mol) 476.47
Formula Weight 476.47
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.