Learn More
Tenofovir hydrate, MedChemExpress
Product Code. 16455908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16455908 10 mg 10mg
16442057 50 mg 50mg
16428547 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16455908 Supplier Medchem Express Supplier No. HY13910A
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Tenofovir hydrate is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.
Tenofovir hydrate is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.
Specifications
Chemical Name or Material Tenofovir hydrate
CAS 206184-49-8
Color White
Purity Grade Notes Research
Molecular Formula C9H16N5O5P
Quantity 10 mg
Synonym GS 1278 hydratePMPA hydrate
Solubility Information DMSO : ≥ 6 mg/mL (19.66 mM)
SMILES C[C@@H](OCP(O)(O)=O)CN1C=NC2=C(N)N=CN=C12.O
Molecular Weight (g/mol) 305.23
Formula Weight 305.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only