Learn More
Tenovin-3, MedChemExpress
Product Code. 16490198
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490198 5 mg 5mg
16473636 25 mg 25mg
16471799 1 mg 1mg
16467738 10 mg 10mg
16403717 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16490198 Supplier Medchem Express Supplier No. HY19339
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
MedChemExpress Tenovin-3 is a p53 activator.
Tenovin-3 is a p53 activator.
Specifications
Chemical Name or Material Tenovin-3
Physical Form Solid
CAS 1011301-27-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H21N3OS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 31 mg/mL (94.67 mM)
SMILES O=C(NC(NC1=CC=C(N)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
Molecular Weight (g/mol) 327.44
Formula Weight 327.44
Percent Purity 99.86%
Grade Research
Show More Show Less
Research purposes only