Learn More
Tenovin-6 Hydrochloride, MedChemExpress
Product Code. 16476786
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476786 100 mg 100mg
16432677 50 mg 50mg
16429157 5 mg 5mg
16426588 10 mg 10mg
16420246 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16476786 Supplier Medchem Express Supplier No. HY15510B
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Tenovin-6 Hydrochloride, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 Hydrochloride inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 Hydrochloride also inhibits dihydroorotate dehydrogenase (DHODH).
Tenovin-6 Hydrochloride, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 Hydrochloride inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 Hydrochloride also inhibits dihydroorotate dehydrogenase (DHODH).
Specifications
Chemical Name or Material Tenovin-6 Hydrochloride
Physical Form Solid
CAS 1011301-29-3
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H35ClN4O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 49 mg/mL (99.78 mM)
SMILES O=C(NC(NC1=CC=C(NC(CCCCN(C)C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2.Cl
Molecular Weight (g/mol) 491.09
Formula Weight 491.09
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only