missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TENR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20598
This item is not returnable.
View return policy
Description
TENR Polyclonal specifically detects TENR in Human samples. It is validated for Western Blot.
Specifications
| TENR | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| adenosine deaminase domain containing 1 (testis-specific), adenosine deaminase domain-containing protein 1, FLJ32741, TENR, Testis nuclear RNA-binding protein | |
| Rabbit | |
| 64 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96M93 | |
| ADAD1 | |
| Recombinant protein encompassing a sequence within the center region of human TENR. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 132612 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction