Learn More
Invitrogen™ Tensin 4 Recombinant Rabbit Monoclonal Antibody (23GB4225), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552667
 View more versions of this product
Product Code. 30089677
  • € 186.00 / 20µL
Description
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tensin 4
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q4V8I3, Q8BZ33, Q8IZW8
TNS4
A synthesized peptide derived from TNS4 (200-233AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB4225
Unconjugated
TNS4
9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4
Rabbit
Affinity chromatography
RUO
217169, 303517, 84951
-20°C