missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teplizumab Antibody (2505B) [DyLight 350], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10245D
This item is not returnable.
View return policy
Description
Teplizumab Monoclonal antibody specifically detects Teplizumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Teplizumab | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Full-length Teplizumab | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 2505B | |
| Western Blot, Flow Cytometry | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction