missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TEPP 46
Description
TEPP 46 is a potent and selective allosteric activator of pyruvate kinase M2 (PKM2) (EC50 = 92 nM); it exhibits no effect on PKM1. TEPP 46 binds at the dimer-dimer interface of the PKM2 homotetramer, blocks its translocation into the nuclues and promotes a constitutively PKM2 active state. TEPP 46 impairs H1299 lung cancer cell proliferation under hypoxic conditions but not under normoxia in vitro. It inhibits growth of H1299 tumor xenografts in mice. TEPP 46 can reverse fructose-derived F1P inhibition of PKM2 and prevent fructose promoted hypoxia adaptation of intestinal cells. TEPP 46 reduces activation, proliferation, and cytokine production of both T helper 17 (Th17) and Th1 cells in vitro. It also inhibits experimental autoimmune encephalomyelitis (EAE) in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 6-[(3-Aminophenyl)methyl]-4,6-dihydro-4-methyl-2-(methylsulfinyl)-5H-thieno[2',3':4,5]pyrrolo[2,3-d]pyridazin-5-one |
| CAS | 1221186-53-3 |
| Quantity | 50 mg |
| Target | PKM2 |
| Molecular Formula | C17H16N4O2S2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?