missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teprotumumab Antibody (975547) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB96341M
This item is not returnable.
View return policy
Description
Teprotumumab Monoclonal antibody specifically detects Teprotumumab in Broad Species Reactivity samples. It is validated for Flow Cytometry, Binding Inhibition
Specifications
| Teprotumumab | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Full-length Teprotumumab | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Binding Inhibition Assay | |
| 975547 | |
| Flow Cytometry, Binding Inhibition | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction