missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119 Antibody (TER-119), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1125PEATT594
This item is not returnable.
View return policy
Description
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry
Specifications
| TER-119 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 104231 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| TER-119 | |
| Flow Cytometry | |
| Ly76, Ly-76, TER119 | |
| C57BL/6 mouse day-14 fetal liver cells | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction