missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TER-119 Monoclonal Antibody (TER-119), Brilliant Violet™ 421, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
€ 147.00 - € 337.00
Specifications
| Antigen | TER-119 |
|---|---|
| Clone | TER-119 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17886961
|
Invitrogen™
404-5921-80 |
25 μg |
€ 147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17862030
|
Invitrogen™
404-5921-82 |
100 μg |
€ 337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.Specifications
| TER-119 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Ly76; Ly-76; TER119 | |
| Ly76 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ly76 |
| TER-119 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Rat | |
| Mouse | |
| 0 | |
| 104231 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title